Assay Details
| Gene Symbol: DNAJC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190204.1 | NP_001177133.1 | 5 - 6 | 432 | 81 | |
| GenBank mRNA | AY610302.1 | - | 5 - 6 | 374 | 81 | |
| EV991770.2 | - | 4 - 5 | 297 | 81 | ||
More Information
Set Membership: |
|

DNAJC19
Genomic Map