Assay Details
Gene Symbol: SNRPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190213.1 | NP_001177142.1 | 3 - 4 | 312 | 83 | |
GenBank mRNA | AY610259.1 | - | 3 - 4 | 286 | 83 | |
BX922649.2 | - | 3 - 4 | 311 | 83 | ||
EV898445.2 | - | 3 - 4 | 345 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
