Assay Details
Gene Symbol: ARPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190239.1 | NP_001177168.1 | 7 - 8 | 613 | 58 | |
GenBank mRNA | AK239011.1 | - | 7 - 8 | 677 | 58 | |
AK399338.1 | - | 7 - 8 | 633 | 58 | ||
AK400742.1 | - | 7 - 8 | 689 | 58 | ||
AY609460.1 | - | 7 - 8 | 613 | 58 | ||
EU868835.1 | - | 6 - 7 | 544 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
