Assay Details
| Gene Symbol: CALB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194980.2 | NP_001181909.2 | 2 - 3 | 242 | 65 | |
| GenBank mRNA | HM627268.1 | - | 2 - 3 | 224 | 65 | |
| HX225950.1 | - | 2 - 3 | 242 | 65 | ||
More Information
Set Membership: |
|

CALB2
Genomic Map