Assay Details
| Gene Symbol: PDIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195112.1 | NP_001182041.1 | 3 - 4 | 370 | 62 | |
| GenBank mRNA | AB282745.1 | - | 3 - 4 | 370 | 62 | |
| AK345882.1 | - | 3 - 4 | 468 | 62 | ||
| AK398695.1 | - | 3 - 4 | 525 | 62 | ||
More Information
Set Membership: |
|

PDIA3
Genomic Map