Assay Details
| Gene Symbol: ACRV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195320.1 | NP_001182249.1 | 2 - 3 | 619 | 84 | |
| XM_021102001.1 | XP_020957660.1 | 428 | 84 | |||
| XM_021102002.1 | XP_020957661.1 | 377 | 84 | |||
| GenBank mRNA | GU373674.1 | - | 2 - 3 | 619 | 84 | |
More Information
Set Membership: |
|

ACRV1
Genomic Map