Assay Details
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195352.1 | NP_001182281.1 | 3 - 4 | 305 | 64 | |
XM_005660938.3 | XP_005660995.1 | 705 | 64 | |||
XM_005660939.3 | XP_005660996.1 | 456 | 64 | |||
XM_005660940.2 | XP_005660997.1 | 469 | 64 | |||
GenBank mRNA | AK350068.1 | - | 3 - 4 | 466 | 64 | |
AY610386.1 | - | 3 - 4 | 276 | 64 | ||
GU373711.1 | - | 3 - 4 | 305 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
