Assay Details
| Gene Symbol: CLIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198923.2 | NP_001185852.2 | 1 - 2 | 423 | 60 | |
| GenBank mRNA | AK395655.1 | - | 1 - 2 | 395 | 60 | |
| AK400732.1 | - | 1 - 2 | 422 | 60 | ||
| EU715290.1 | - | 1 - 2 | 352 | 60 | ||
More Information
Set Membership: |
|

CLIC5
Genomic Map