Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204852.1 | NP_001191781.1 | 1 - 2 | 145 | 88 | |
| GenBank mRNA | AK230541.1 | - | 1 - 2 | 102 | 88 | |
| FD599314.1 | - | 1 - 2 | 137 | 88 | ||
More Information
Set Membership: |
|

FPGT
Genomic Map