Assay Details
| Gene Symbol: IL1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302388.2 | NP_001289317.2 | 4 - 5 | 526 | 103 | |
| Gene Symbol: LOC110258125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081283.1 | XP_020936942.1 | 299 | 103 | ||
| Gene Symbol: LOC110258579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081829.1 | XP_020937488.1 | 296 | 103 | ||
| Gene Symbol: LOC110259156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082463.1 | XP_020938122.1 | 296 | 103 | ||
More Information
Set Membership: |
|

IL1B2
Genomic Map