Assay Details
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097465.1 | NP_001090934.1 | 1 - 2 | 74 | 76 | |
GenBank mRNA | AK344769.1 | - | 1 - 2 | 109 | 76 | |
AY609707.2 | - | 178 | 76 | |||
DQ629140.1 | - | 1 - 2 | 74 | 76 | ||
DQ629141.1 | - | 1 - 2 | 74 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
