Assay Details
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134353.2 | NP_001127825.1 | 1 - 2 | 311 | 95 | |
GenBank mRNA | AK235116.1 | - | 1 - 2 | 331 | 95 | |
EF095722.2 | - | 1 - 2 | 311 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
