Assay Details
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134353.2 | NP_001127825.1 | 3 - 4 | 1008 | 93 | |
GenBank mRNA | AK235116.1 | - | 3 - 4 | 1028 | 93 | |
EF095722.2 | - | 3 - 4 | 1008 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
