Assay Details
| Gene Symbol: SLC30A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139475.1 | NP_001132947.1 | 2 - 3 | 496 | 75 | |
| GenBank mRNA | EU825192.2 | - | 2 - 3 | 496 | 75 | |
More Information
Set Membership: |
|

SLC30A2
Genomic Map