Assay Details
Gene Symbol: LGALS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142843.1 | NP_001136315.1 | 1 - 2 | 28 | 52 | |
XM_021093249.1 | XP_020948908.1 | 779 | 52 | |||
XM_021093250.1 | XP_020948909.1 | 1188 | 52 | |||
GenBank mRNA | FJ424705.1 | - | 1 - 2 | 28 | 52 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142843.1, XM_021093249.1, XM_021093250.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
