Assay Details
| Gene Symbol: MGLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143718.1 | NP_001137190.1 | 7 - 8 | 881 | 79 | |
| XM_013982013.2 | XP_013837467.1 | 968 | 79 | |||
| XM_013982014.2 | XP_013837468.2 | 1321 | 79 | |||
| XM_021068336.1 | XP_020923995.1 | 1011 | 79 | |||
| GenBank mRNA | AK350271.1 | - | 5 - 6 | 623 | 79 | |
| AK352498.1 | - | 7 - 8 | 896 | 79 | ||
| AK399967.1 | - | 7 - 8 | 898 | 79 | ||
| FJ436393.1 | - | 7 - 8 | 881 | 79 | ||
More Information
Set Membership: |
|

MGLL
Genomic Map