Assay Details
Gene Symbol: CLDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161645.1 | NP_001155117.1 | 1 | 626 | 62 | |
XM_005662194.3 | XP_005662251.1 | 1163 | 62 | |||
GenBank mRNA | FJ873113.1 | - | 1 | 626 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
