Assay Details
Gene Symbol: CDIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167642.1 | NP_001161114.1 | 3 - 4 | 327 | 106 | |
XM_005655088.3 | XP_005655145.1 | 734 | 106 | |||
XM_021085630.1 | XP_020941289.1 | 718 | 106 | |||
GenBank mRNA | AB529449.1 | - | 3 - 4 | 327 | 106 | |
AK236165.1 | - | 3 - 4 | 663 | 106 | ||
AK236320.1 | - | 3 - 4 | 680 | 106 | ||
AK351493.1 | - | 3 - 4 | 670 | 106 | ||
AK393252.1 | - | 3 - 4 | 740 | 106 | ||
AK399176.1 | - | 3 - 4 | 672 | 106 | ||
AK400901.1 | - | 3 - 4 | 702 | 106 | ||
GU144288.1 | - | 3 - 4 | 638 | 106 | ||
GU144289.1 | - | 3 - 4 | 647 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
