Assay Details
| Gene Symbol: EBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167646.1 | NP_001161118.1 | 3 - 4 | 466 | 70 | |
| XM_005657789.1 | XP_005657846.1 | 706 | 70 | |||
| XM_005657790.2 | XP_005657847.1 | 769 | 70 | |||
| XM_005657791.3 | XP_005657848.1 | 744 | 70 | |||
| XM_013990872.2 | XP_013846326.1 | 708 | 70 | |||
| GenBank mRNA | AB529458.1 | - | 3 - 4 | 466 | 70 | |
| AK232804.1 | - | 4 - 5 | 638 | 70 | ||
| AK236024.1 | - | 5 - 6 | 731 | 70 | ||
| AK236559.1 | - | 4 - 5 | 862 | 70 | ||
| AK389357.1 | - | 4 - 5 | 651 | 70 | ||
| AY609764.1 | - | 4 - 5 | 642 | 70 | ||
More Information
Set Membership: |
|

EBP
Genomic Map