Assay Details
Gene Symbol: ATG9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190280.1 | NP_001177209.1 | 4 - 5 | 837 | 72 | |
XM_021078467.1 | XP_020934126.1 | 1226 | 72 | |||
XM_021078468.1 | XP_020934127.1 | 1230 | 72 | |||
GenBank mRNA | HM046519.1 | - | 4 - 5 | 837 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
