Assay Details
| Gene Symbol: BAMBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201485.1 | NP_001188414.1 | 1 - 2 | 377 | 74 | |
| GenBank mRNA | AK231676.1 | - | 1 - 2 | 405 | 74 | |
| AK347849.1 | - | 1 - 2 | 400 | 74 | ||
| HQ009517.1 | - | 1 - 2 | 377 | 74 | ||
More Information
Set Membership: |
|

BAMBI
Genomic Map