Assay Details
| Gene Symbol: BHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204771.1 | NP_001191700.1 | 5 - 6 | 612 | 56 | |
| GenBank mRNA | HQ130334.1 | - | 5 - 6 | 612 | 56 | |
More Information
Set Membership: |
|

BHMT2
Genomic Map