Assay Details
| Gene Symbol: UBE2V1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205094.1 | NP_001192023.1 | 1 - 2 | 88 | 86 | |
| GenBank mRNA | AK238798.1 | - | 1 - 2 | 88 | 86 | |
| AK350493.1 | - | 1 - 2 | 88 | 86 | ||
| AK351450.1 | - | 1 - 2 | 66 | 86 | ||
| AK391410.1 | - | 1 - 2 | 46 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205094.1 transcripts.
Set Membership: |
|

UBE2V1
Genomic Map