Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206375.1 | NP_001193304.1 | 2 - 3 | 156 | 113 | |
XM_005653167.2 | XP_005653224.1 | 259 | 113 | |||
XM_021091042.1 | XP_020946701.1 | 522 | 113 | |||
GenBank mRNA | EW465602.2 | - | 2 - 3 | 163 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
