Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206375.1 | NP_001193304.1 | 6 - 7 | 730 | 66 | |
XM_005653167.2 | XP_005653224.1 | 833 | 66 | |||
XM_021091042.1 | XP_020946701.1 | 1096 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
