Assay Details
| Gene Symbol: ATP5MC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244862.1 | NP_001231791.1 | 2 - 3 | 166 | 94 | |
| GenBank mRNA | AK232718.1 | - | 2 - 3 | 184 | 94 | |
| AK344759.1 | - | 2 - 3 | 162 | 94 | ||
| AK348215.1 | - | 2 - 3 | 162 | 94 | ||
| GQ913653.1 | - | 1 - 2 | 108 | 94 | ||
More Information
Set Membership: |
|

ATP5MC3
Genomic Map