Assay Details
| Gene Symbol: RPLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244866.1 | NP_001231795.1 | 3 - 4 | 236 | 67 | |
| GenBank mRNA | AK239881.1 | - | 3 - 4 | 232 | 67 | |
| AK345249.1 | - | 3 - 4 | 232 | 67 | ||
| AK346519.1 | - | 3 - 4 | 234 | 67 | ||
| AK347730.1 | - | 3 - 4 | 236 | 67 | ||
| AK350260.1 | - | 3 - 4 | 232 | 67 | ||
| AK352241.1 | - | 3 - 4 | 232 | 67 | ||
| AK391916.1 | - | 3 - 4 | 233 | 67 | ||
More Information
Set Membership: |
|

RPLP2
Genomic Map