Assay Details
| Gene Symbol: GRPEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244870.1 | NP_001231799.1 | 2 - 3 | 231 | 65 | |
| XM_021078518.1 | XP_020934177.1 | 780 | 65 | |||
| XM_021078521.1 | XP_020934180.1 | 254 | 65 | |||
| GenBank mRNA | CF176583.1 | - | 2 - 3 | 231 | 65 | |
More Information
Set Membership: |
|

GRPEL2
Genomic Map