Assay Details
| Gene Symbol: PLA2G2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244874.1 | NP_001231803.1 | 2 - 3 | 274 | 77 | |
| GenBank mRNA | AK349876.1 | - | 2 - 3 | 276 | 77 | |
More Information
Set Membership: |
|

PLA2G2F
Genomic Map