Assay Details
Gene Symbol: MARCHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244915.1 | NP_001231844.1 | 4 - 5 | 732 | 67 | |
GenBank mRNA | AK236835.1 | - | 4 - 5 | 732 | 67 | |
AK396059.1 | - | 4 - 5 | 738 | 67 | ||
AY609952.1 | - | 4 - 5 | 732 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
