Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244917.1 | NP_001231846.1 | 2 - 3 | 153 | 93 | |
| XM_005665239.3 | XP_005665296.2 | 263 | 93 | |||
| GenBank mRNA | AK231118.1 | - | 2 - 3 | 157 | 93 | |
| AK236342.1 | - | 2 - 3 | 153 | 93 | ||
| AK344747.1 | - | 2 - 3 | 128 | 93 | ||
| AY610032.1 | - | 2 - 3 | 129 | 93 | ||
More Information
Set Membership: |
|

MYCBP
Genomic Map