Assay Details
| Gene Symbol: SUMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244932.1 | NP_001231861.1 | 4 - 5 | 620 | 56 | |
| XM_013981911.2 | XP_013837365.2 | 456 | 56 | |||
| GenBank mRNA | AK351541.1 | - | 4 - 5 | 620 | 56 | |
| AK389451.1 | - | 4 - 5 | 628 | 56 | ||
| AK395786.1 | - | 4 - 5 | 624 | 56 | ||
| AK400781.1 | - | 4 - 5 | 620 | 56 | ||
More Information
Set Membership: |
|

SUMF1
Genomic Map