Assay Details
| Gene Symbol: TACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244958.1 | NP_001231887.1 | 1 - 2 | 479 | 81 | |
| GenBank mRNA | AK231964.1 | - | 1 - 2 | 485 | 81 | |
| AK234409.1 | - | 1 - 2 | 479 | 81 | ||
| AK398120.1 | - | 1 - 2 | 494 | 81 | ||
More Information
Set Membership: |
|

TACO1
Genomic Map