Assay Details
| Gene Symbol: TACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244958.1 | NP_001231887.1 | 4 - 5 | 888 | 89 | |
| XM_005668679.3 | XP_005668736.1 | 1068 | 89 | |||
| GenBank mRNA | AK231964.1 | - | 4 - 5 | 894 | 89 | |
| AK234409.1 | - | 4 - 5 | 888 | 89 | ||
| AK398120.1 | - | 4 - 5 | 903 | 89 | ||
More Information
Set Membership: |
|

TACO1
Genomic Map