Assay Details
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244966.1 | NP_001231895.1 | 9 - 10 | 738 | 61 | |
XM_021066107.1 | XP_020921766.1 | 857 | 61 | |||
XM_021066108.1 | XP_020921767.1 | 821 | 61 | |||
XM_021066109.1 | XP_020921768.1 | 690 | 61 | |||
GenBank mRNA | AK235599.1 | - | 9 - 10 | 738 | 61 | |
AK389245.1 | - | 1948 | 61 | |||
AK395099.1 | - | 2025 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
