Assay Details
| Gene Symbol: CAPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244975.1 | NP_001231904.1 | 3 - 4 | 371 | 82 | |
| NM_001244976.1 | NP_001231905.1 | 3 - 4 | 346 | 82 | ||
| XM_005661298.3 | XP_005661355.1 | 430 | 82 | |||
| XM_005661299.3 | XP_005661356.1 | 430 | 82 | |||
| GenBank mRNA | AK231677.1 | - | 3 - 4 | 346 | 82 | |
| AK234487.1 | - | 3 - 4 | 371 | 82 | ||
| AK351024.1 | - | 3 - 4 | 346 | 82 | ||
More Information
Set Membership: |
|

CAPS
Genomic Map