Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244979.1 | NP_001231908.1 | 1 - 2 | 203 | 62 | |
| GenBank mRNA | AK344461.1 | - | 1 - 2 | 203 | 62 | |
| AK396695.1 | - | 1 - 2 | 288 | 62 | ||
More Information
Set Membership: |
|

ACOT13
Genomic Map