Assay Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245001.1 | NP_001231930.1 | 5 - 6 | 1024 | 75 | |
XM_021088657.1 | XP_020944316.1 | 2076 | 75 | |||
GenBank mRNA | AK343398.1 | - | 5 - 6 | 1024 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
