Assay Details
| Gene Symbol: ZNF232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245009.1 | NP_001231938.1 | 2 - 3 | 933 | 86 | |
| XM_005669150.3 | XP_005669207.1 | 1025 | 86 | |||
| XM_005669152.3 | XP_005669209.1 | 566 | 86 | |||
| XM_021066111.1 | XP_020921770.1 | 1105 | 86 | |||
| GenBank mRNA | AK349548.1 | - | 2 - 3 | 933 | 86 | |
More Information
Set Membership: |
|

ZNF232
Genomic Map