Assay Details
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246203.1 | NP_001233132.1 | 4 - 5 | 286 | 68 | |
XM_005673561.3 | XP_005673618.1 | 2336 | 68 | |||
GenBank mRNA | HQ266638.1 | - | 4 - 5 | 286 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
