Assay Details
Gene Symbol: GPR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251818.1 | NP_001238747.1 | 1 | 1124 | 61 | |
GenBank mRNA | HM777011.1 | - | 1 | 1124 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
