Assay Details
| Gene Symbol: BIVM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252098.1 | NP_001239027.1 | 9 - 10 | 1438 | 91 | |
| XM_021065131.1 | XP_020920790.1 | 1614 | 91 | |||
| XM_021065132.1 | XP_020920791.1 | 1835 | 91 | |||
| XM_021065133.1 | XP_020920792.1 | 1629 | 91 | |||
| XM_021065134.1 | XP_020920793.1 | 1423 | 91 | |||
| XM_021065135.1 | XP_020920794.1 | 1341 | 91 | |||
| GenBank mRNA | AK232672.1 | - | 9 - 10 | 1215 | 91 | |
| AK393584.1 | - | 9 - 10 | 1709 | 91 | ||
| CN161877.1 | - | 5 - 6 | 514 | 91 | ||
More Information
Set Membership: |
|

BIVM
Genomic Map