Assay Details
| Gene Symbol: PRSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206438.1 | NP_001193367.1 | 3 - 4 | 460 | 107 | |
| GenBank mRNA | BX920898.2 | - | 3 - 4 | 447 | 107 | |
| BX924569.2 | - | 3 - 4 | 463 | 107 | ||
More Information
Set Membership: |
|

PRSS2
Genomic Map