Assay Details
| Gene Symbol: METTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206443.1 | NP_001193372.1 | 1 - 2 | 1212 | 78 | |
| NM_001206445.1 | NP_001193374.1 | 1 - 2 | 1212 | 78 | ||
| NM_001206446.1 | NP_001193375.1 | 1 - 2 | 1212 | 78 | ||
| XM_013981369.2 | XP_013836823.1 | 1225 | 78 | |||
| XM_021068433.1 | XP_020924092.1 | 459 | 78 | |||
| XM_021068434.1 | XP_020924093.1 | 1193 | 78 | |||
| GenBank mRNA | AK233563.1 | - | 1 - 2 | 1211 | 78 | |
| AK345156.1 | - | 1 - 2 | 1212 | 78 | ||
More Information
Set Membership: |
|

METTL6
Genomic Map