Assay Details
| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243218.1 | NP_001230147.1 | 6 - 7 | 644 | 118 | |
| GenBank mRNA | AK239991.1 | - | 6 - 7 | 640 | 118 | |
| BP170171.1 | - | 6 - 7 | 645 | 118 | ||
More Information
Set Membership: |
|

EIF1AX
Genomic Map