Assay Details
| Gene Symbol: UPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243289.1 | NP_001230218.1 | 2 - 3 | 551 | 87 | |
| XM_021079380.1 | XP_020935039.1 | 606 | 87 | |||
| GenBank mRNA | AK343621.1 | - | 2 - 3 | 551 | 87 | |
More Information
Set Membership: |
|

UPRT
Genomic Map