Assay Details
| Gene Symbol: CST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243310.2 | NP_001230239.1 | 3 - 4 | 390 | 78 | |
| GenBank mRNA | AJ938954.1 | - | 3 - 4 | 391 | 78 | |
| AK399795.1 | - | 3 - 4 | 471 | 78 | ||
More Information
Set Membership: |
|

CST7
Genomic Map