Assay Details
Gene Symbol: SPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243315.1 | NP_001230244.1 | 2 - 3 | 923 | 83 | |
GenBank mRNA | AK349141.1 | - | 2 - 3 | 923 | 83 | |
AK400606.1 | - | 2 - 3 | 924 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
