Assay Details
Gene Symbol: LRRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243334.1 | NP_001230263.1 | 10 - 11 | 1249 | 58 | |
XM_005662841.3 | XP_005662898.1 | 2262 | 58 | |||
XM_021088516.1 | XP_020944175.1 | 2117 | 58 | |||
GenBank mRNA | AK349579.1 | - | 9 - 10 | 1025 | 58 | |
AW314215.1 | - | 1 - 2 | 111 | 58 | ||
DY435962.1 | - | 2 - 3 | 456 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
