Assay Details
| Gene Symbol: RPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243345.1 | NP_001230274.1 | 3 - 4 | 328 | 63 | |
| XM_013994186.2 | XP_013849640.1 | 484 | 63 | |||
| GenBank mRNA | AK231738.1 | - | 2 - 3 | 486 | 63 | |
| AK237333.1 | - | 3 - 4 | 301 | 63 | ||
| AK347687.1 | - | 3 - 4 | 294 | 63 | ||
| AK350211.1 | - | 2 - 3 | 481 | 63 | ||
| BP171914.1 | - | 3 - 4 | 331 | 63 | ||
More Information
Set Membership: |
|

RPL13
Genomic Map